Assay Details
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081933.1 | NP_001075402.1 | 4 - 5 | 313 | 73 | |
| GenBank mRNA | AJ006515.1 | - | 2 - 3 | 182 | 73 | |
| AJ582012.1 | - | 4 - 5 | 313 | 73 | ||
More Information
Set Membership: |
|

BSP1
Genomic Map