Assay Details
| Gene Symbol: MHCB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082508.2 | NP_001075977.1 | 2 - 3 | 342 | 57 | |
| GenBank mRNA | X80018.1 | - | 342 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MHCB3| Gene Symbol: MHCB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082508.2 | NP_001075977.1 | 2 - 3 | 342 | 57 | |
| GenBank mRNA | X80018.1 | - | 342 | 57 | ||
Set Membership: |
|