Assay Details
| Gene Symbol: CD68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099762.1 | NP_001093232.1 | 1 - 2 | 103 | 106 | |
| GenBank mRNA | AY373259.2 | - | 1 - 2 | 103 | 106 | |
| AY499346.1 | - | 1 - 2 | 62 | 106 | ||
| JL615906.1 | - | 1 - 2 | 128 | 106 | ||
More Information
Set Membership: |
|

CD68
Genomic Map