Assay Details
Gene Symbol: RSPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109682.1 | NP_001103152.1 | 2 - 3 | 295 | 140 | |
GenBank mRNA | EU029662.1 | - | 2 - 3 | 295 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() |
