Assay Details
| Gene Symbol: MILL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036560.1 | 4 - 5 | 914 | 80 | ||
| GenBank mRNA | AB126064.1 | - | 4 - 5 | 914 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MILL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036560.1 | 4 - 5 | 914 | 80 | ||
| GenBank mRNA | AB126064.1 | - | 4 - 5 | 914 | 80 | |
Set Membership: |
|