Assay Details
Gene Symbol: MILL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036560.1 | 6 - 7 | 1084 | 92 | ||
GenBank mRNA | AB126064.1 | - | 6 - 7 | 1084 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
