Assay Details
Gene Symbol: DRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142815.1 | NP_001136287.1 | 1 - 2 | 175 | 78 | |
GenBank mRNA | JQ254089.1 | - | 1 - 2 | 78 | ||
JQ254090.1 | - | 1 - 2 | 130 | 78 | ||
JQ254091.1 | - | 1 - 2 | 130 | 78 | ||
JQ254092.1 | - | 1 - 2 | 130 | 78 | ||
JQ254094.1 | - | 1 - 2 | 130 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
