Assay Details
Gene Symbol: DRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142815.1 | NP_001136287.1 | 4 - 5 | 838 | 82 | |
GenBank mRNA | JQ254090.1 | - | 4 - 5 | 793 | 82 | |
JQ254092.1 | - | 4 - 5 | 793 | 82 | ||
JQ254093.1 | - | 4 - 5 | 793 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
