Assay Details
| Gene Symbol: MIP-2BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ469975.1 | - | 3 - 4 | 327 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIP-2BETA| Gene Symbol: MIP-2BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ469975.1 | - | 3 - 4 | 327 | 95 | |
Set Membership: |
|