Assay Details
| Gene Symbol: ACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081838.1 | NP_001075307.1 | 1 | 67 | 108 | |
| XM_023655002.1 | XP_023510770.1 | 205 | 108 | |||
| GenBank mRNA | AF035774.1 | - | 1 | 67 | 108 | |
More Information
Set Membership: |
|

ACTB
Genomic Map