Assay Details
Gene Symbol: MIR652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033110.1 | 1 | 44 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC111771635 | LOC111771635 | -215.4kb | Sense | Chr. X - 90749845 - 90754310 on NCBI Build EquCab3.0 | |
Downstream | AMMECR nuclear protein 1 | AMMECR1 | 127.8kb | Anti-sense | Chr. X - 91097572 - 91205033 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
