Assay Details
| Gene Symbol: RPLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085436.1 | NP_001078905.1 | 1 - 2 | 127 | 78 | |
| XM_023653768.1 | XP_023509536.1 | 78 | ||||
| GenBank mRNA | AY246717.1 | - | 1 - 2 | 127 | 78 | |
More Information
Set Membership: |
|

RPLP2
Genomic Map