Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163880.1 | NP_001157352.1 | 3 - 4 | 225 | 98 | |
| XM_005601758.2 | XP_005601815.1 | 460 | 98 | |||
More Information
Set Membership: |
|

RPL35A
Genomic Map