Assay Details
Gene Symbol: RNASE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163987.1 | NP_001157459.1 | 1 - 2 | 36 | 75 | |
GenBank mRNA | AJ580632.1 | - | 1 - 2 | 36 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ580632.1, NM_001163987.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
