Assay Details
| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166039.2 | NP_001159511.1 | 1 - 2 | 358 | 78 | |
| GenBank mRNA | GQ429291.1 | - | 1 - 2 | 334 | 78 | |
More Information
Set Membership: |
|

FASLG
Genomic Map