Assay Details
| Gene Symbol: HNRNPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242509.1 | NP_001229438.1 | 1 - 2 | 120 | 77 | |
| XM_014740750.2 | XP_014596236.1 | 151 | 77 | |||
| XM_014740752.2 | XP_014596238.1 | 151 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001242509.1, XM_014740750.2, XM_014740752.2 transcripts.
Set Membership: |
|

HNRNPA1
Genomic Map