Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170338.1 | NP_001163809.1 | 1 - 2 | 356 | 58 | |
| GenBank mRNA | CD464773.1 | - | 1 - 2 | 92 | 58 | |
| CD536845.1 | - | 1 - 2 | 81 | 58 | ||
| CX600675.1 | - | 1 - 2 | 357 | 58 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map