Assay Details
| Gene Symbol: TMED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202561.1 | NP_001189490.1 | 2 - 3 | 637 | 84 | |
| GenBank mRNA | JL619886.1 | - | 2 - 3 | 283 | 84 | |
More Information
Set Membership: |
|

TMED7
Genomic Map