Assay Details
Gene Symbol: TICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202854.1 | NP_001189783.1 | 3 - 4 | 216 | 87 | |
GenBank mRNA | CD536283.1 | - | 3 - 4 | 216 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001202854.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
