Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204743.1 | NP_001191672.1 | 3 - 4 | 350 | 110 | |
| XM_023651653.1 | XP_023507421.1 | 515 | 110 | |||
| GenBank mRNA | DN511018.1 | - | 4 - 5 | 399 | 110 | |
More Information
Set Membership: |
|

NME2
Genomic Map