Assay Details
Gene Symbol: MIR135A-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033061.1 | 1 | 52 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NEDD1 gamma-tubulin ring complex targeting factor | NEDD1 | -567.6kb | Sense | Chr. 28 - 22699064 - 22745172 on NCBI Build EquCab3.0 | |
Downstream | thymopoietin | TMPO | 721.4kb | Sense | Chr. 28 - 24034303 - 24062576 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
