Assay Details
Gene Symbol: MIR92B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032848.1 | 1 | 35 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thrombospondin 3 | THBS3 | -344.0 | Anti-sense | Chr. 5 - 39305934 - 39317012 on NCBI Build EquCab3.0 | |
Downstream |
mucin 1, cell surface associated |
MUC1 | 1.6kb | Anti-sense | Chr. 5 - 39319003 - 39323892 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
