Assay Details
Gene Symbol: LOC100055155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606631.3 | XP_005606688.1 | 867 | 62 | ||
XM_014736832.2 | XP_014592318.1 | 1072 | 62 | |||
XM_014736833.2 | XP_014592319.1 | 769 | 62 | |||
XM_014736834.2 | XP_014592320.1 | 1532 | 62 | |||
XM_023631663.1 | XP_023487431.1 | 887 | 62 | |||
XM_023631666.1 | XP_023487434.1 | 887 | 62 | |||
XM_023631667.1 | XP_023487435.1 | 874 | 62 | |||
XM_023631668.1 | XP_023487436.1 | 887 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
