Assay Details
Gene Symbol: MIR21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032904.1 | 1 | 32 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peptidyl-tRNA hydrolase 2 | PTRH2 | -124.4kb | Anti-sense | Chr. 11 - 34027411 - 34036968 on NCBI Build EquCab3.0 | |
Downstream | tubulin delta 1 | TUBD1 | 13.4kb | Anti-sense | Chr. 11 - 34174800 - 34199043 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
