Assay Details
Gene Symbol: MIR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032929.1 | 1 | 37 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | minichromosome maintenance complex component 7 | MCM7 | Sense | Chr. 13 - 8114061 - 8121909 on NCBI Build EquCab3.0 | ||
Downstream | microRNA mir-93 | MIR93 | eca-mir-93 | 136.0 | Sense | Chr. 13 - 8115133 - 8115191 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
