Assay Details
Gene Symbol: MIR885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032956.1 | 1 | 31 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 6 member 11 | SLC6A11 | -392.3kb | Anti-sense | Chr. 16 - 7543963 - 7665492 on NCBI Build EquCab3.0 | |
Downstream |
SEC13 homolog, nuclear pore and COPII coat complex component |
SEC13 | 70.0kb | Sense | Chr. 16 - 8127821 - 8156432 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
