Assay Details
Gene Symbol: MIR1296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032777.1 | 1 | 53 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | jumonji domain containing 1C | JMJD1C | Sense | Chr. 1 - 52677730 - 53023906 on NCBI Build EquCab3.0 | ||
Downstream | receptor accessory protein 3 | REEP3 | 176.7kb | Anti-sense | Chr. 1 - 53023396 - 53119761 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
