Assay Details
Gene Symbol: MIR7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032791.1 | 1 | 37 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | apoptosis enhancing nuclease | AEN | -6.6kb | Sense | Chr. 1 - 95470219 - 95481312 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-1179 | MIR1179 | eca-mir-1179 | 3.2kb | Sense | Chr. 1 - 95491143 - 95491228 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
