Assay Details
Gene Symbol: MIR200B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032796.1 | 1 | 29 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-3548 | MIR3548 | eca-mir-3548 | -555.0 | Anti-sense | Chr. 2 - 48713785 - 48713913 on NCBI Build EquCab3.0 |
Downstream | chromosome 2 C1orf159 homolog | C2H1orf159 | 52.8kb | Anti-sense | Chr. 2 - 48767368 - 48795853 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
