Assay Details
Gene Symbol: MIR429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032805.1 | 1 | 33 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tubulin tyrosine ligase like 10 | TTLL10 | -3.3kb | Sense | Chr. 2 - 48692902 - 48709161 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-3548 | MIR3548 | eca-mir-3548 | 1.3kb | Anti-sense | Chr. 2 - 48713785 - 48713913 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
