Assay Details
Gene Symbol: MIR761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032809.1 | 1 | 36 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RAB3B, member RAS oncogene family |
RAB3B | -69.8kb | Sense | Chr. 2 - 7313734 - 7373122 on NCBI Build EquCab3.0 | |
Downstream | oxysterol binding protein like 9 | OSBPL9 | 59.1kb | Anti-sense | Chr. 2 - 7502029 - 7658057 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
