Assay Details
Gene Symbol: MIR206-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032973.1 | 1 | 29 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PKHD1 ciliary IPT domain containing fibrocystin/polyductin | PKHD1 | -46.7kb | Anti-sense | Chr. 20 - 50373816 - 50790646 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-133b | MIR133B | eca-mir-133b | 4.0kb | Sense | Chr. 20 - 50841419 - 50841479 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
