Assay Details
Gene Symbol: MIR219-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032974.1 | 1 | 42 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxysteroid 17-beta dehydrogenase 8 | HSD17B8 | -997.0 | Sense | Chr. 20 - 34555673 - 34557892 on NCBI Build EquCab3.0 | |
Downstream | ring finger protein 1 | RING1 | 834.0 | Sense | Chr. 20 - 34559809 - 34563426 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
