Assay Details
Gene Symbol: MIR103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032982.1 | 1 | 41 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ring finger protein 24 | RNF24 | -8.1kb | Anti-sense | Chr. 22 - 19399241 - 19471796 on NCBI Build EquCab3.0 | |
Downstream | mitochondrial antiviral signaling protein | MAVS | 39.3kb | Sense | Chr. 22 - 19519257 - 19536004 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
