Assay Details
Gene Symbol: MIRLET7D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032987.1 | 1 | 44 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | putative serine protease 47 | LOC111770229 | -131.8kb | Sense | Chr. 23 - 53814999 - 53826019 on NCBI Build EquCab3.0 | |
Downstream | microRNA let-7f | MIRLET7F | eca-let-7f | 1.9kb | Sense | Chr. 23 - 53959741 - 53959827 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
