Assay Details
Gene Symbol: MIRLET7F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032988.1 | 1 | 48 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | basic proline-rich protein-like | LOC111770145 | Anti-sense | Chr. 23 - 53953848 - 53968726 on NCBI Build EquCab3.0 | ||
Downstream | protein tyrosine phosphatase domain containing 1 | PTPDC1 | 39.6kb | Sense | Chr. 23 - 53999434 - 54070775 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
