Assay Details
Gene Symbol: MIR154A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033007.1 | 1 | 30 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-485 | MIR485 | eca-mir-485 | -4.1kb | Sense | Chr. 24 - 43364618 - 43364690 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-154b | MIR154B | eca-mir-154b | 187.0 | Sense | Chr. 24 - 43369045 - 43369185 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
