Assay Details
Gene Symbol: MIR337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033011.1 | 1 | 45 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-493b | MIR493B | eca-mir-493b | -10.2kb | Sense | Chr. 24 - 43167633 - 43167721 on NCBI Build EquCab3.0 |
Downstream | retrotransposon Gag like 1 | RTL1 | 4.9kb | Anti-sense | Chr. 24 - 43182999 - 43192234 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
