Assay Details
| Gene Symbol: MIR345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033013.1 | 1 | 39 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 25 member 29 | SLC25A29 | -1.5kb | Anti-sense | Chr. 24 - 42642735 - 42656043 on NCBI Build EquCab3.0 | |
| Downstream | solute carrier family 25 member 47 | SLC25A47 | 17.5kb | Sense | Chr. 24 - 42675127 - 42683267 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR345
Genomic Map