Assay Details
Gene Symbol: MIR369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033014.1 | 1 | 30 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-412 | MIR412 | eca-mir-412 | -70.0 | Sense | Chr. 24 - 43374416 - 43374495 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-410 | MIR410 | eca-mir-410 | 262.0 | Sense | Chr. 24 - 43374896 - 43374969 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
