Assay Details
Gene Symbol: MIR187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032880.1 | 1 | 29 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | polypeptide N-acetylgalactosaminyltransferase 1 | GALNT1 | -138.0kb | Anti-sense | Chr. 8 - 59299140 - 59373257 on NCBI Build EquCab3.0 | |
Downstream | chromosome 8 C18orf21 homolog | C8H18orf21 | 39.0kb | Anti-sense | Chr. 8 - 59550371 - 59558737 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
