Assay Details
Gene Symbol: MIR301B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032881.1 | 1 | 53 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | stromal cell derived factor 2 like 1 | SDF2L1 | -3.6kb | Sense | Chr. 8 - 5377765 - 5379858 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-130b | MIR130B | eca-mir-130b | 235.0 | Sense | Chr. 8 - 5383817 - 5383897 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
