Assay Details
Gene Symbol: MIR151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032884.1 | 1 | 28 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | argonaute RISC catalytic component 2 | AGO2 | EIF2C2 | -112.9kb | Sense | Chr. 9 - 81788239 - 81870567 on NCBI Build EquCab3.0 |
Downstream | DENN domain containing 3 | DENND3 | 308.7kb | Anti-sense | Chr. 9 - 82292177 - 82352954 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
