Assay Details
Gene Symbol: MIR150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032890.1 | 1 | 27 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribosomal protein S11 | RPS11 | -1.3kb | Anti-sense | Chr. 10 - 19588976 - 19591159 on NCBI Build EquCab3.0 | |
Downstream | Fc fragment of IgG receptor and transporter | FCGRT | 8.1kb | Anti-sense | Chr. 10 - 19600622 - 19612929 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
