Assay Details
Gene Symbol: MIR1180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032897.1 | 1 | 32 | 54 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | serine protease 33-like | LOC102149409 | -12.8kb | Sense | Chr. 11 - 59564978 - 59567719 on NCBI Build EquCab3.0 | |
Downstream | epsin 2 | EPN2 | 6.8kb | Anti-sense | Chr. 11 - 59587340 - 59666438 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
