Assay Details
Gene Symbol: MIR196A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032903.1 | 1 | 31 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox B9 | HOXB9 | -6.0kb | Sense | Chr. 11 - 24744060 - 24749411 on NCBI Build EquCab3.0 | |
Downstream | homeobox B13 | HOXB13 | 59.0kb | Sense | Chr. 11 - 24814491 - 24816779 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
