Assay Details
Gene Symbol: MIR632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032916.1 | 1 | 33 | 54 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
proteasome 26S subunit, non-ATPase 11 |
PSMD11 | -71.5kb | Sense | Chr. 11 - 40367036 - 40396432 on NCBI Build EquCab3.0 | |
Downstream | chromosome 11 C17orf75 homolog | C11H17orf75 | 87.0 | Anti-sense | Chr. 11 - 40468072 - 40482677 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
