Assay Details
Gene Symbol: MIR493A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032922.1 | 1 | 35 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synaptotagmin 13 | SYT13 | -407.8kb | Sense | Chr. 12 - 9967843 - 10011352 on NCBI Build EquCab3.0 | |
Downstream | carbohydrate sulfotransferase 1 | CHST1 | 17.9kb | Sense | Chr. 12 - 10437050 - 10536855 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
