Assay Details
Gene Symbol: MIR493B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033034.1 | 1 | 45 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-770 | MIR770 | eca-mir-770 | -17.2kb | Sense | Chr. 24 - 43150327 - 43150421 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-337 | MIR337 | eca-mir-337 | 10.2kb | Sense | Chr. 24 - 43177970 - 43178061 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
