Assay Details
Gene Symbol: MIR541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033039.1 | 1 | 41 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-377 | MIR377 | eca-mir-377 | -1.9kb | Sense | Chr. 24 - 43371371 - 43371439 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-409 | MIR409 | eca-mir-409 | 837.0 | Sense | Chr. 24 - 43374266 - 43374344 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
